6-Bromo-2,4-dichloropyrido[2,3-d]pyrimidine ≥95%
Supplier: Ambeed
|
Warning
|
A941394-1G
A941394-250MG
77573-876EA
151.41
USD
77573-876
77686-969
6-Bromo-2,4-dichloropyrido[2,3-d]pyrimidine ≥95%
6-Bromo-2,4-dichloropyrido[2,3-d]pyrimidine
Precaución: for Research Use Only
Formula:
C₇H₂BrCl₂N₃ MW: 278.92 g/mol |
MDL Number:
MFCD17015892 CAS Number: 1234616-70-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...