3-Cyano-1-propylboronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)butanenitrile

A941232-1G A941232-250MG A941232-5G
77891-706EA 78 USD
77891-706 77891-707 77891-708
3-Cyano-1-propylboronic acid pinacol ester ≥98%
3-Cyano-1-propylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₀H₁₈BNO₂
MDL Number: MFCD09953480
CAS Number: 238088-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR