Decyl decanoate ≥98%

Supplier: Ambeed
Warning

A941068-25G A941068-100G A941068-5G A941068-500G
77603-326EA 15.38 USD
77603-326 77670-374 77670-376 77880-762
Decyl decanoate ≥98%
Decyl decanoate

Precaución: for Research Use Only

Formula: C₂₀H₄₀O₂
MDL Number: MFCD00059280
CAS Number: 1654-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR