N-{[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}acetamide ≥95%

Supplier: Ambeed
Warning

A938415-100MG A938415-250MG A938415-5G
77599-278EA 17.43 USD
77599-278 77623-172 77784-388
N-{[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}acetamide ≥95%
N-{[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}acetamide

Precaución: for Research Use Only

Formula: C₁₅H₂₂BNO₃
MDL Number: MFCD18383571
CAS Number: 1235450-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR