2'-Deoxyguanosine 5'-triphosphate ≥98%

Supplier: Ambeed
Warning

Synonyms: dGTP

A938182-10MG A938182-1MG A938182-5MG
77618-594EA 69.01 USD
77618-594 77618-596 77618-598
2'-Deoxyguanosine 5'-triphosphate ≥98%
2'-Deoxyguanosine 5'-triphosphate

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₅O₁₃P₃
MW: 507.18 g/mol
MDL Number: MFCD01632476
CAS Number: 2564-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR