(S)-2-Amino-4-(4-hydroxyphenyl)butanoic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: H-HoTyr-OH·HBr

A937415-100MG A937415-1G A937415-250MG
77595-518EA 130.18 USD
77595-518 77595-520 77595-522
(S)-2-Amino-4-(4-hydroxyphenyl)butanoic acid ≥95%
(S)-2-Amino-4-(4-hydroxyphenyl)butanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.22 g/mol
MDL Number: MFCD03490505
CAS Number: 221243-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR