tert-Butyl-3-amino-3-methylpyrrolidine-1-carboxylate ≥98%

Supplier: Ambeed
Warning

A937327-250MG A937327-100MG A937327-5G
77674-400EA 96 USD
77674-400 77884-882 77884-883
tert-Butyl-3-amino-3-methylpyrrolidine-1-carboxylate ≥98%
tert-Butyl-3-amino-3-methylpyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD12408544
CAS Number: 1158758-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR