2-(4-Bromo-2-fluoro-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-fluoro-5-(trifluoromethyl)phenylboronic acid, pinacol ester

A936442-1G A936442-250MG A936442-5G
77784-384EA 217 USD
77784-384 77784-385 77784-386
2-(4-Bromo-2-fluoro-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98%
2-(4-Bromo-2-fluoro-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₄BBrF₄O₂
MDL Number: MFCD20441906
CAS Number: 2096337-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR