2-Methyl-2H-pyrazole-3-sulfonyl chloride ≥98%

Supplier: Ambeed
Danger

Synonyms: 1-Methyl-1H-pyrazole-5-sulphonyl chloride

A935450-1G A935450-100MG A935450-250MG
77603-320EA 189.63 USD
77603-320 77674-394 77674-396
2-Methyl-2H-pyrazole-3-sulfonyl chloride ≥98%
2-Methyl-2H-pyrazole-3-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₄H₅ClN₂O₂S
MW: 180.61 g/mol
MDL Number: MFCD09065021
CAS Number: 1020721-61-2
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR