(E)-Methyl 3-(4-hydroxy-3-methoxyphenyl)acrylate ≥98%

Supplier: Ambeed
Warning

Synonyms: (E)-Ferulic acid methyl ester

A934727-25G A934727-5G
77670-366EA 285 USD
77670-366 77670-368
(E)-Methyl 3-(4-hydroxy-3-methoxyphenyl)acrylate ≥98%
(E)-Methyl 3-(4-hydroxy-3-methoxyphenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
MDL Number: MFCD00017208
CAS Number: 22329-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR