Fmoc-3-chloro-L-homophenylalanine ≥95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-3-Cl-HoPhe-OH

A933664-100MG A933664-250MG A933664-1G
77784-378EA 163 USD
77784-378 77784-380 77784-379
Fmoc-3-chloro-L-homophenylalanine ≥95%
Fmoc-3-chloro-L-homophenylalanine

Precaución: for Research Use Only

Formula: C₂₅H₂₂ClNO₄
MDL Number: MFCD07372175
CAS Number: 1260587-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR