5-Bromo-1-methylpyrazole-4-carbonitrile ≥95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1-methyl-1H-pyrazole-4-carbonitrile

A932093-250MG A932093-1G
77573-864EA 19.57 USD
77573-864 77674-388
5-Bromo-1-methylpyrazole-4-carbonitrile ≥95%
5-Bromo-1-methylpyrazole-4-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₄BrN₃
MW: 186.009 g/mol
MDL Number: MFCD17168487
CAS Number: 1269293-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR