3'-Methoxy-2,2,2-trifluoroacetophenone ≥98%

Supplier: Ambeed
Warning

A929791-5G A929791-1G
77674-386EA 137 USD
77674-386 77884-870
3'-Methoxy-2,2,2-trifluoroacetophenone ≥98%
3'-Methoxy-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
MDL Number: MFCD00461895
CAS Number: 30724-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR