Thieno[3,2-c]pyridine-2-carbaldehyde ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A929789-100MG A929789-250MG
77614-650EA 32.96 USD
77614-650 77684-881
Thieno[3,2-c]pyridine-2-carbaldehyde ≥98%, Ambeed.Inc
Thieno[3,2-c]pyridine-2-carbaldehyde

Precaución: For research use only.

Formula: C₈H₅NOS
MDL Number: MFCD18453108
CAS Number: 94226-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR