2-(2-Fluoroethoxy)ethanol ≥97%

Supplier: Ambeed
Warning

A928750-1G A928750-250MG
77623-160EA 282.22 USD
77623-160 77784-375
2-(2-Fluoroethoxy)ethanol ≥97%
2-(2-Fluoroethoxy)ethanol

Precaución: for Research Use Only

Formula: C₄H₉FO₂
MDL Number: MFCD01727880
CAS Number: 373-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR