(8-Oxo-5,6,7,8-tetrahydronaphthalen-2-yl)boronic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: (8-Oxo-6,7-dihydro-5H-naphthalen-2-yl)boronic acid

A928551-100MG A928551-250MG A928551-1G A928551-5G
77595-502EA 96.35 USD
77595-502 77595-504 77686-962 77901-463
(8-Oxo-5,6,7,8-tetrahydronaphthalen-2-yl)boronic acid ≥97%
(8-Oxo-5,6,7,8-tetrahydronaphthalen-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BO₃
MDL Number: MFCD28122670
CAS Number: 1135871-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR