Sodium 6,6'-(ethene-1,2-diyl)bis(3-((4-morpholino-6-(phenylamino)-1,3,5-triazin-2-yl)amino)benzenesulfonate) ≥97%

Supplier: Ambeed
Warning

A928377-25G A928377-5G
77880-753EA 47 USD
77880-753 77880-754
Sodium 6,6'-(ethene-1,2-diyl)bis(3-((4-morpholino-6-(phenylamino)-1,3,5-triazin-2-yl)amino)benzenesulfonate) ≥97%
Sodium 6,6'-(ethene-1,2-diyl)bis(3-((4-morpholino-6-(phenylamino)-1,3,5-triazin-2-yl)amino)benzenesulfonate)

Precaución: for Research Use Only

Formula: C₄₀H₃₈N₁₂Na₂O₈S₂
MDL Number: MFCD00071913
CAS Number: 16090-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR