Fmoc-trans-4-fluoro-Pro-Oh ≥98%

Supplier: Ambeed
Warning

Synonyms: (2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-fluoropyrrolidine-2-carboxylic acid, (2S,4R)-4-Fluoropyrrolidine-2-carboxylic acid, N-FMOC protected, Fmoc-trans-4-F-Pro-OH

A926988-1G A926988-250MG A926988-100MG A926988-5G
77573-854EA 95 USD
77573-854 77595-498 77603-310 77882-830
Fmoc-trans-4-fluoro-Pro-Oh ≥98%
Fmoc-trans-4-fluoro-Pro-Oh

Precaución: for Research Use Only

Formula: C₂₀H₁₈FNO₄
MDL Number: MFCD03424224
CAS Number: 203866-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR