1,3-Dibromo-2,4-dimethylbenzene ≥96%, Ambeed.Inc

Supplier: Ambeed
Warning

A926337-5G A926337-25G
77684-879EA 57 USD
77684-879 77891-694
1,3-Dibromo-2,4-dimethylbenzene ≥96%, Ambeed.Inc
1,3-Dibromo-2,4-dimethylbenzene

Precaución: For research use only.

Formula: C₈H₈Br₂
MDL Number: MFCD11846009
CAS Number: 90434-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR