3-Acetyl-6-bromo-2H-chromen-2-one ≥98%

Supplier: Ambeed
Warning

A925773-100MG A925773-1G A925773-5G A925773-250MG
77670-358EA 18 USD
77670-358 77670-360 77670-362 77880-752
3-Acetyl-6-bromo-2H-chromen-2-one ≥98%
3-Acetyl-6-bromo-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₁₁H₇BrO₃
MDL Number: MFCD00024075
CAS Number: 2199-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR