NDSB-195 ≥98%

Supplier: Ambeed
Danger

Synonyms: Ethyl(dimethylaminio)propane sulphonate, 3-(Ethyldimethylaminio)propane-1-sulphonate, 3-(Ethyldimethylammonio)propane-1-sulphonate, NDSB 195, Dimethylethylammonium propane sulphonate

A925197-1G A925197-5G
77674-378EA 44 USD
77674-378 77674-380
NDSB-195 ≥98%
NDSB-195

Precaución: for Research Use Only

Formula: C₇H₁₇NO₃S
MW: 195.277 g/mol
MDL Number: MFCD01319124
CAS Number: 160255-06-1
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR