7-(Diethylamino)-3-(thiophene-3-carbonyl)-2H-chromen-2-one ≥97%

Supplier: Ambeed
Warning

A924956-1G A924956-5G
77686-960EA 86 USD
77686-960 77686-961
7-(Diethylamino)-3-(thiophene-3-carbonyl)-2H-chromen-2-one ≥97%
7-(Diethylamino)-3-(thiophene-3-carbonyl)-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₁₈H₁₇NO₃S
MDL Number: MFCD00051376
CAS Number: 77820-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR