(2-Phenylpyridin-3-yl)boronic acid ≥96%

Supplier: Ambeed
Warning

Synonyms: 2-Phenylpyridine-3-boronic acid

A923817-100MG A923817-1G A923817-250MG
77886-913EA 107 USD
77886-913 77886-914 77886-915
(2-Phenylpyridin-3-yl)boronic acid ≥96%
(2-Phenylpyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀BNO₂
MW: 199.01 g/mol
MDL Number: MFCD07438014
CAS Number: 1029654-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR