2-Isopropoxy-1,3-diisopropylbenzene ≥95%

Supplier: Ambeed
Warning

A922840-100MG A922840-1G
77884-864EA 32 USD
77884-864 77884-865
2-Isopropoxy-1,3-diisopropylbenzene ≥95%
2-Isopropoxy-1,3-diisopropylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₂₄O
MDL Number: MFCD21363770
CAS Number: 141214-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR