(E)-((2R,3R,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-4,4-difluoro-3-hydroxytetrahydrofuran-2-yl)methyl octadec-9-enoate ≥96%

Supplier: Ambeed
Warning

Synonyms: Gemcitabine elaidate

A922777-1MG A922777-10MG A922777-5MG
77618-570EA 20.6 USD
77618-570 77747-684 77747-685
(E)-((2R,3R,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-4,4-difluoro-3-hydroxytetrahydrofuran-2-yl)methyl octadec-9-enoate ≥96%
(E)-((2R,3R,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-4,4-difluoro-3-hydroxytetrahydrofuran-2-yl)methyl octadec-9-enoate

Precaución: for Research Use Only

Formula: C₂₇H₄₃F₂N₃O₅
MW: 527.64 g/mol
MDL Number: MFCD15145214
CAS Number: 210829-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR