2-(Chloromethyl)-3,5-difluoropyridine ≥98%

Supplier: Ambeed
Danger

A922691-100MG A922691-1G
77893-728EA 76 USD
77893-728 77893-729
2-(Chloromethyl)-3,5-difluoropyridine ≥98%
2-(Chloromethyl)-3,5-difluoropyridine

Precaución: for Research Use Only

Formula: C₆H₄ClF₂N
MDL Number: MFCD11520701
CAS Number: 1222633-85-3
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR