Quinazolin-8-amine ≥95%

Supplier: Ambeed
Warning

A922548-100MG A922548-250MG
77880-749EA 160 USD
77880-749 77880-750
Quinazolin-8-amine ≥95%
Quinazolin-8-amine

Precaución: for Research Use Only

Formula: C₈H₇N₃
MW: 145.16 g/mol
MDL Number: MFCD09029725
CAS Number: 101421-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR