Cobalt tetrafluoroborate hexahydrate ≥98%

Supplier: Ambeed
Danger

A922345-5G A922345-25G
77573-848EA 13 USD
77573-848 77900-793
Cobalt tetrafluoroborate hexahydrate ≥98%
Cobalt tetrafluoroborate hexahydrate

Precaución: for Research Use Only

Formula: B₂CoF₈H₁₂O₆
MDL Number: MFCD00243274
CAS Number: 15684-35-2
UN: 3260
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR