N-(2-(3-(Piperazin-1-ylmethyl)imidazo[2,1-b]thiazol-6-yl)phenyl)quinoxaline-2-carboxamide ≥99%

Supplier: Ambeed
Warning

Synonyms: SRT 1720

A921472-10MG A921472-5MG
77674-370EA 83 USD
77674-370 77674-372
N-(2-(3-(Piperazin-1-ylmethyl)imidazo[2,1-b]thiazol-6-yl)phenyl)quinoxaline-2-carboxamide ≥99%
N-(2-(3-(Piperazin-1-ylmethyl)imidazo[2,1-b]thiazol-6-yl)phenyl)quinoxaline-2-carboxamide

Precaución: for Research Use Only

Formula: C₂₅H₂₃N₇OS
MW: 469.56 g/mol
MDL Number: MFCD14584468
CAS Number: 925434-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR