N-Methyl-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetamide ≥95%

Supplier: Ambeed
Warning

A921311-1G A921311-250MG A921311-5G A921311-100MG
77599-266EA 126.08 USD
77599-266 77599-268 77618-564 77749-746
N-Methyl-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetamide ≥95%
N-Methyl-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetamide

Precaución: for Research Use Only

Formula: C₁₂H₂₀BN₃O₃
MW: 265.12 g/mol
MDL Number: MFCD21337769
CAS Number: 1346245-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR