(S)-Ethyl 3-aminobutanoate hydrochloride ≥93%

Supplier: Ambeed
Warning

A920874-5G A920874-1G
77618-562EA 247.2 USD
77618-562 77749-745
(S)-Ethyl 3-aminobutanoate hydrochloride ≥93%
(S)-Ethyl 3-aminobutanoate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO₂
MW: 167.63 g/mol
MDL Number: MFCD01862144
CAS Number: 187876-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR