5-Chloro-6-morpholinopyridine-3-boronic acid ≥98%
Supplier: Ambeed
|
Warning
|
A920519-1G
A920519-5G
A920519-250MG
77784-357EA
209
USD
77784-357
77784-359
77784-358
5-Chloro-6-morpholinopyridine-3-boronic acid ≥98%
5-Chloro-6-morpholinopyridine-3-boronic acid
Precaución: for Research Use Only
Formula:
C₉H₁₂BClN₂O₃ MW: 242.47 g/mol |
MDL Number:
MFCD22684696 CAS Number: 2130896-19-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...