2-Bromo-5-(difluoromethyl)thiophene ≥95%

Supplier: Ambeed
Warning

A920244-250MG A920244-5G
77623-148EA 37.08 USD
77623-148 77784-356
2-Bromo-5-(difluoromethyl)thiophene ≥95%
2-Bromo-5-(difluoromethyl)thiophene

Precaución: for Research Use Only

Formula: C₅H₃BrF₂S
MDL Number: MFCD22378823
CAS Number: 924646-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR