Cyclopropyl(trimethylsilyl)acetylene ≥95%

Supplier: Ambeed
Danger

Synonyms: (Cyclopropylethynyl)trimethylsilane

A919849-1G A919849-25G A919849-10G
77627-756EA 16.09 USD
77627-756 77627-758 77880-747
Cyclopropyl(trimethylsilyl)acetylene ≥95%
Cyclopropyl(trimethylsilyl)acetylene

Precaución: for Research Use Only

Formula: C₈H₁₄Si
MDL Number: MFCD00671355
CAS Number: 81166-84-9
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR