tert-Butyl ((3R,4R)-4-hydroxypyrrolidin-3-yl)carbamate ≥97%

Supplier: Ambeed
Warning

A918685-100MG A918685-250MG
77595-478EA 73.8 USD
77595-478 77618-560
tert-Butyl ((3R,4R)-4-hydroxypyrrolidin-3-yl)carbamate ≥97%
tert-Butyl ((3R,4R)-4-hydroxypyrrolidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₃
MW: 202.25 g/mol
MDL Number: MFCD22399082
CAS Number: 1203566-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR