Methyl 2-(1H-imidazol-5-yl)acetate ≥97%
Supplier: Ambeed
|
Warning
|
A917933-1G
A917933-250MG
A917933-5G
77891-685EA
215
USD
77891-685
77891-686
77891-687
Methyl 2-(1H-imidazol-5-yl)acetate ≥97%
Methyl 2-(1H-imidazol-5-yl)acetate
Precaución: for Research Use Only
Formula:
C₆H₈N₂O₂ |
MDL Number:
MFCD00266723 CAS Number: 4200-46-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...