2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)chromenylium chloride ≥98%

Supplier: Ambeed
Warning

Synonyms: Kuromanin chloride

A917560-100MG A917560-250MG A917560-50MG A917560-5MG
77670-348EA 755 USD
77670-348 77670-350 77670-352 77670-354
2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)chromenylium chloride ≥98%
2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)chromenylium chloride

Precaución: for Research Use Only

Formula: C₂₁H₂₁ClO₁₁
MW: 484.84 g/mol
MDL Number: MFCD27665410
CAS Number: 7084-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR