CCT-137690 ≥98%

Supplier: Ambeed
Warning

Synonyms: 3-((4-(6-Bromo-2-(4-(4-methylpiperazin-1-yl)phenyl)-3H-imidazo[4,5-b]pyridin-7-yl)piperazin-1-yl)methyl)-5-methylisoxazole, CCT 137690, 3-[[4-[6-bromo-2-[4-(4-methylpiperazin-1-yl)phenyl]-1H-imidazo[4,5-b]pyridin-7-yl]piperazin-1-yl]methyl]-5-methyl-1,2-oxazole

A917251-1MG A917251-5MG
77618-558EA 33.99 USD
77618-558 77747-682
CCT-137690 ≥98%
CCT-137690

Precaución: for Research Use Only

Formula: C₂₆H₃₁BrN₈O
MW: 551.48 g/mol
MDL Number: MFCD18206879
CAS Number: 1095382-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR