1-(4-Amino-1H-pyrazol-1-yl)-2-methylpropan-2-ol ≥98%

Supplier: Ambeed
Warning

A915442-1G A915442-250MG
77749-736EA 90 USD
77749-736 77749-737
1-(4-Amino-1H-pyrazol-1-yl)-2-methylpropan-2-ol ≥98%
1-(4-Amino-1H-pyrazol-1-yl)-2-methylpropan-2-ol

Precaución: for Research Use Only

Formula: C₇H₁₃N₃O
MW: 155.2 g/mol
MDL Number: MFCD20703139
CAS Number: 1206640-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR