N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
DABCYL acid, SE, DABCYL N-Succinimidyl Ester, 4-[4-(Dimethylamino)phenylazo]benzoic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl 4-((4-(dimethylamino)phenyl)diazenyl)benzoate
A914566-1G
A914566-250MG
A914566-100MG
77618-554EA
335.78
USD
77618-554
77749-735
77749-734
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate ≥97%
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate
Precaución: for Research Use Only
Formula:
C₁₉H₁₈N₄O₄ MW: 366.38 g/mol |
MDL Number:
MFCD00467415 CAS Number: 146998-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...