tert-Butyl 4-bromobenzyl(methyl)carbamate ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (4-bromobenzyl)(methyl)carbamate

A914553-100MG A914553-1G A914553-250MG
77684-867EA 41 USD
77684-867 77684-868 77684-869
tert-Butyl 4-bromobenzyl(methyl)carbamate ≥95%, Ambeed.Inc
tert-Butyl 4-bromobenzyl(methyl)carbamate

Precaución: For research use only.

Formula: C₁₃H₁₈BrNO₂
MW: 300.19 g/mol
MDL Number: MFCD09701229
CAS Number: 260809-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR