9,10-Dimethoxy-5,6-dihydro-[1,3]dioxolo[4,5-g]isoquinolino[3,2-a]isoquinolin-7-ium hydrogensulfate 99+%

Supplier: Ambeed
Warning

Synonyms: Berberine sulphate

A914013-1G A914013-25G A914013-5G A914013-100G A914013-1MG A914013-250MG
77603-278EA 13.33 USD
77603-278 77603-280 77603-282 77627-740 77627-742 77627-744
9,10-Dimethoxy-5,6-dihydro-[1,3]dioxolo[4,5-g]isoquinolino[3,2-a]isoquinolin-7-ium hydrogensulfate 99+%
9,10-Dimethoxy-5,6-dihydro-[1,3]dioxolo[4,5-g]isoquinolino[3,2-a]isoquinolin-7-ium hydrogensulfate

Precaución: for Research Use Only

Formula: C₂₀H₁₉NO₈S
MW: 433.43 g/mol
MDL Number: MFCD00075789
CAS Number: 633-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR