5-Phenylpentan-1-amine ≥95%

Supplier: Ambeed
Danger

A913925-100MG A913925-5G A913925-1G A913925-250MG
77618-548EA 40.23 USD
77618-548 77749-732 77749-730 77749-731
5-Phenylpentan-1-amine ≥95%
5-Phenylpentan-1-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₇N
MDL Number: MFCD00870521
CAS Number: 17734-21-3
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR