4-Methoxy-2-(trifluoromethyl)phenylboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxy-2-(trifluoromethyl)benzeneboronic acid, (4-Methoxy-2-(trifluoromethyl)phenyl)boronic acid

A913448-1G A913448-250MG
77674-354EA 27 USD
77674-354 77674-356
4-Methoxy-2-(trifluoromethyl)phenylboronic acid ≥98%
4-Methoxy-2-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₃
MW: 219.95 g/mol
MDL Number: MFCD03095346
CAS Number: 313546-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR