2-Methoxy-3-(methoxycarbonyl)phenylboronic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: (2-Methoxy-3-(methoxycarbonyl)phenyl)boronic acid

A913377-1G A913377-250MG A913377-100MG A913377-5G
77595-470EA 165.03 USD
77595-470 77599-258 77623-136 77784-348
2-Methoxy-3-(methoxycarbonyl)phenylboronic acid ≥97%
2-Methoxy-3-(methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₅
MW: 209.992 g/mol
MDL Number: MFCD23379502
CAS Number: 1332637-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR