Xantphos Palladacycle Gen. 2 ≥98%

Supplier: Ambeed
Danger

Synonyms: Chloro[(4,5-bis(diphenylphosphino)-9,9-dimethylxanthene)-2-(2-amino-1,1-biphenyl)]palladium(ii)

A911963-5G A911963-1G A911963-250MG
77573-828EA 108 USD
77573-828 77686-947 77686-948
Xantphos Palladacycle Gen. 2 ≥98%
Xantphos Palladacycle Gen. 2

Precaución: for Research Use Only

Formula: C₅₁H₄₂ClNOP₂Pd
MDL Number: MFCD22666442
CAS Number: 1375325-77-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR