Methyl-2-methylthiazole-4-carboxylate ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A911303-1G A911303-5G
77684-866EA 17 USD
77684-866 77886-907
Methyl-2-methylthiazole-4-carboxylate ≥98%, Ambeed.Inc
Methyl-2-methylthiazole-4-carboxylate

Precaución: For research use only.

Formula: C₆H₇NO₂S
MDL Number: MFCD09870034
CAS Number: 6436-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR