1,1,1-Trifluoro-2-butanone ≥95%

Supplier: Ambeed
Danger

A910599-25G A910599-5G
77573-824EA 101 USD
77573-824 77747-676
1,1,1-Trifluoro-2-butanone ≥95%
1,1,1-Trifluoro-2-butanone

Precaución: for Research Use Only

Formula: C₄H₅F₃O
MDL Number: MFCD00190641
CAS Number: 381-88-4
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR