(S)-8,8-Dimethyl-2-oxo-7,8-dihydro-2H,6H-pyrano[3,2-g]chromen-7-yl 3-methylbut-2-enoate ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Decursin
A909799-1MG
A909799-10MG
A909799-5MG
77599-254EA
47.15
USD
77599-254
77627-732
77627-734
(S)-8,8-Dimethyl-2-oxo-7,8-dihydro-2H,6H-pyrano[3,2-g]chromen-7-yl 3-methylbut-2-enoate ≥98%
(S)-8,8-Dimethyl-2-oxo-7,8-dihydro-2H,6H-pyrano[3,2-g]chromen-7-yl 3-methylbut-2-enoate
Precaución: for Research Use Only
Formula:
C₁₉H₂₀O₅ MW: 328.36 g/mol |
MDL Number:
MFCD00272154 CAS Number: 5928-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...