Methyl 3-aminobicyclo[1.1.1]pentane-1-carboxylate hydrochloride ≥95%

Supplier: Ambeed
Warning

A909488-250MG A909488-100MG A909488-1G A909488-25G A909488-5G
77614-626EA 51.5 USD
77614-626 77686-946 77893-707 77893-708 77893-709
Methyl 3-aminobicyclo[1.1.1]pentane-1-carboxylate hydrochloride ≥95%
Methyl 3-aminobicyclo[1.1.1]pentane-1-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₁NO₂·ClH
MDL Number: MFCD20720060
CAS Number: 676371-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR